Learn More
CPC Scientific Ac-Cys-Glu-His-DPhe-Arg-Trp-Cys-Lys-Pro-Val-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MSHS005B
Encompass
A superagonist with ultraprolonged melanotropic activity in frog and lizard skin bioassays. It exhibits a complete resistance to tryptic degradation. ONE-LETTER SEQUENCE: Ac-CEH-DPhe-RWCKPV-NH2 (Cys1 and 7 bridge)MOLECULAR FORMULA: C61H86N18O13S2MOLECULAR WEIGHT:1343.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [91050-39-4], SYNONYMS: melanotropinsRESEARCH AREA: HormonalREFERENCES: W.L. Cody et al., J. Med. Chem., 28, 683 (1985)
Catalog No. 50-210-9842
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.