Learn More
Enzo Life Sciences Ac-DEVD-AFC (1mg). CAS: 201608-14-2
SDP
Supplier:  Enzo Life Sciences ALX260032M001
Encompass_Preferred
Fluorogenic substrate for caspase-3 (CPP32), with a Km=9.7µM) and related cysteine proteases. Sequence is based on PARP cleavage at Asp216 for caspase-3. Similar to Ac-DEVD-AMC (ALX-260-031) but the AFC fluorophore has a greater Stokes’ shift upon cleavage than AMC. Reaction can be monitored quantitatively or visually using a hand-held long-UV lamp and visualizing a blue to green shift in fluorescence upon cleavage. Ex.: 400nm, Em.: 505nm. Alternative name: Caspase-3 substrate (fluorogenic). Formula: C30H34F3N5O13. MW: 729.6. Purity: ≥96% (HPLC). Appearance: White to off-white powder. Peptide content: 65-95%. Solubility: Soluble in dimethyl formamide, DMSO or methanol; slightly soluble in water (0.4mg/ml). Long Term Storage: -20°C. Handling: Protect from light. Keep cool and dry.
Catalog No. 50-200-8243
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.