Learn More
STEMCELL Technologies Ac-DEVD-CHO (Trifluoroacetate Salt), Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 1000537
Encompass_Preferred
Reversibly inhibit caspase-3 and caspase-7 with Ac-DEVD-CHO (Garcia-Calvo et al. J Biol Chem 1998 Talanian et al. J Biol Chem 1997). Ac-DEVD-CHO is a caspase inhibitor with the PARP cleavage site DEVD which blocks PARP cleavage activity and inhibits caspase-3 and caspase-7 in a reversible manner (Garcia-Calvo et al. J Biol Chem 1998 Talanian et al. J Biol Chem 1997). This inhibition is via the aldehyde group’s interaction with the active site cysteine of these caspases (Thornberry et al. Perspect Drug Discov Des 1995). Caspase inhibitors are used to investigate caspase activation and apoptosis. Ac-DEVD-CHO is also used in cancer research to block camptothecin-induced apoptosis in bone tumor cells. Ac-DEVD-CHO is supplied as a trifluoroacetate salt with ≥95% purity Molecular weight 502.5 g/mol. Visit STEMCELL.com for more information and related products.
Catalog No. 50-239-0192
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.