Learn More
CPC Scientific Ac-Asp-Glu-Val-Asp-pNA 5MG
SDP
Supplier:  CPC Scientific CASP052B
Encompass
Cleavage of this chromogenic peptide substrate for caspase-3 is detected at a wavelength of 405 nm. ONE-LETTER SEQUENCE: Ac-DEVD-pNAMOLECULAR FORMULA: C26H34N6O13MOLECULAR WEIGHT:638.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [189950-66-1], RESEARCH AREA: ApoptosisREFERENCES: D.W. Nicholson, Nature, 376 (1995)
Catalog No. 50-210-9011
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.