Learn More
Cayman Chemical PDGFR TKI III 5mg
SDP
Supplier:  Cayman Chemical 149255 MG
Encompass_Preferred
Inhibits the uptake of dopamine serotonin and norepinephrine with IC50 values of 0.77 6.8 and 0.42 M respectively indicating potency comparable to cocaine or methamphetamine intended for forensic purposes
Catalog No. 50-297-0399
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.