Learn More
Enzo Life Sciences Ac-DMQD-AMC (5mg)
SDP
Supplier:  Enzo Life Sciences ALX260078M005
Encompass_Preferred
Caspase-3 substrate. Fluorogenic substrate for caspase-3. Alternative name: Caspase 3 substrate (fluorogenic). Formula: C30H38N6O12S. MW: 706.7. Purity: ≥98% (HPLC). Appearance: White powder. Peptide content: 75-95%. Solubility: Soluble in DMSO. Dilute with distilled water or buffer. Also soluble in phosphate buffer, pH 7.5. Long Term Storage: -20°C. Handling: Keep cool and dry.
Catalog No. 50-200-8293
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.