Learn More
CPC Scientific Ac-Asp-Met-Gln-Asp-CHO 1MG
SDP
Supplier:  CPC Scientific CASP054A
Encompass
Specific inhibitor of caspase-3. Together with the inhibitor CASP-072 this peptide is capable of dissecting the pathway of caspase activation in Fas-stimulated Jurkat cells.SEQUENCE: Ac-Asp-Met-Gln-Asp-CHOONE-LETTER SEQUENCE: Ac-DMQD-CHOMOLECULAR FORMULA: C20H31N5O10S1MOLECULAR WEIGHT: 534.54STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [259199-63-8]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:H. Hirata et al., J. Exp. Med., 187, 587 (1998)SKU(s): CASP-054A, CASP-054B
Catalog No. 50-210-9014