Learn More
CPC Scientific Ac-Asp-Thr-Glu-Asp- Val-Val-Pro-Nva-O-4-phenylazophenyl ester 1MG
SDP
Supplier:  CPC Scientific SUBS050B
Encompass
This peptide is a spectrophotometric substrate for a chymotrypsin-like serine protease encoded by a hepatitis C virus (HCV) and is promising target antiviral therapeutic development. Due to the low catalytic efficiency of HCV?s serine protease, this continuous spectrophotometric substrate is a ideal for both kinetic analysis and high-throughput screening. It is hydrolyzed by HCV protease under second-order rate kinetics with a constant (kcat/Km) of 80,000 ? 10,000 M-1s-1.SEQUENCE: Ac-Asp-Thr-Glu-Asp- Val-Val-Pro-Nva-O-4-phenylazophenyl esterONE-LETTER SEQUENCE: Ac-DTEDVVP-Nva-O-4-phenylazophenyl esterMOLECULAR FORMULA: C51H70N10O17MOLECULAR WEIGHT: 1095.2STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and Phosphatases REFERENCES:Zhang R, Beyer BM, Durkin J et al Anal Biochem 270:268-275 (1999)SKU(s): SUBS-050A, SUBS-050B
Catalog No. 50-211-0529
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.