Learn More
Cayman Chemical Ac-ETD-pNA 1mg
SDP
Supplier:  Cayman Chemical 245681 MG
Encompass_Preferred
A caspase-8 substrate; cleavage of the IETD peptide sequence releases p-nitroalinide, which can be quantified by colorimetric detection at 405 nm as a measure of enzyme activity
Catalog No. 50-293-5982