Learn More
Enzo Life Sciences Ac-IETD-CHO (1mg). CAS: 191338-86-0
SDP
Supplier:  Enzo Life Sciences ALX260043M001
Encompass_Preferred
Reversible inhibitor of caspase-6, -8 and granzyme B. Also inhibits caspase-10. Alternative name: Caspase 8 inhibitor (aldehyde), Granzyme B inhibitor (aldehyde). Formula: C21H34N4O10. MW: 502.5. Purity: ≥96% (HPLC). Formulation: Lyophilized. Solubility: Soluble in distilled water at 10 mg/mL and in DMSO at 1 mg/mL. Long Term Storage: -20°C.
Catalog No. 50-200-8253
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.