Learn More
CPC Scientific Ac-Leu-Pro-Phe-Phe-Asp-NH2 0.5MG
Supplier: CPC Scientific AMYD033A
beta-sheet breaker peptide iAbeta5p that crosses the blood-brain barrier at a higher rate than most proteins and peptides. It is known to be selectively taken up by the brain, thereby suggesting that the peptide is being specifically transported to the brain. A significant increase in neuronal survival and decrease in brain inflammation associated with the reduction of amyloid plaques in two different transgenic Alzheimer's disease (AD) models is additionally reported for this pentapeptide. Abeta (1-42) Fibrillogenesis Inhibitor.SEQUENCE: Ac-Leu-Pro-Phe-Phe-Asp-NH2ONE-LETTER SEQUENCE: Ac-LPFFD-NH2MOLECULAR FORMULA: C35H46N6O8MOLECULAR WEIGHT: 678.79STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [339990-02-2]SYNONYMS: Acetyl-(Pro,Asp)-Amyloid -Protein (17-21) amideRESEARCH AREA: Alzheimer's DiseaseREFERENCES:SKU(s): AMYD-033A, AMYD-033B
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.