Learn More
CPC Scientific Ac-(NMe)Tyr-Val-Ala-Asp-CHO 0.5MG
SDP
Supplier:  CPC Scientific CASP002A
Encompass
ONE-LETTER SEQUENCE: Ac-(NMe)Tyr-Val-Ala-Asp-CHOMOLECULAR FORMULA: C24H34N4O8MOLECULAR WEIGHT:506.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [160806-26-8], RESEARCH AREA: ApoptosisREFERENCES: M.D.Mullican et al., Bioorg. Med. Chem. Letters, 4, 2359 (1994)
Catalog No. 50-210-8931
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.