Learn More
CPC Scientific Ac-Val-Ala-Asp-CMK 1MG
SDP
Supplier:  CPC Scientific CASP017A
Encompass
ONE-LETTER SEQUENCE: Ac-VAD-CMKMOLECULAR FORMULA: C15H24N3O6Cl1MOLECULAR WEIGHT:377.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Apoptosis
Catalog No. 50-210-8951
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.