Learn More
CPC Scientific Ac-Val-Asp-Val-Ala-Asp-pNA 5MG
SDP
Supplier:  CPC Scientific CASP044B
Encompass
ONE-LETTER SEQUENCE: Ac-VDVAD-pNAMOLECULAR FORMULA: C29H74N7O12MOLECULAR WEIGHT:679.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [189684-53-5], RESEARCH AREA: ApoptosisREFERENCES: R.V.Talanian et al., J. Biol. Chem. , 272, 9677 (1997)
Catalog No. 50-210-8997
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.