Learn More
CPC Scientific Ac-Val-Asp-Val-Ala-Asp-CHO 5MG
Supplier: CPC Scientific CASP043B
SEQUENCE: Ac-Val-Asp-Val-Ala-Asp-CHOONE-LETTER SEQUENCE: Ac-VDVAD-CHOMOLECULAR FORMULA: C23H37N5O10MOLECULAR WEIGHT: 543.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [194022-51-0]SYNONYMS: Ac-Val-Asp-Val-Ala-Asp-aldehyde (pseudo acid)RESEARCH AREA: ApoptosisREFERENCES:R. V. Talanian et al., J. Biol. Chem., 272, 9677 (1997) SKU(s): CASP-043A, CASP-043B, CASP-043C, CASP-043D
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.