Learn More
Enzo Life Sciences Ac-VEID-AMC (1mg). CAS: 219137-97-0
SDP
Supplier:  Enzo Life Sciences ALX260064M001
Encompass_Preferred
Caspase-6 substrate. Fluorogenic substrate for caspase-6 (Km=30µML, Kcat/Km=168000M-1sec-1) and related cysteine proteases. Sequence is based on lamin A site of cleavage. Ex.:340-360nM, Em.:440-460nm. Alternative name: Caspase 6 substrate (fluorogenic). Formula: C32H43N5O11. MW: 673.7. Purity: ≥95% (HPLC). Peptide content: 70-90%. Solubility: Soluble in DMSO; dilute with distilled water or buffer; also soluble in phosphate buffer, pH 7.5. Long Term Storage: -20°C. Handling: Protect from light. Keep cool and dry.
Catalog No. 50-200-8279