Learn More
CPC Scientific Ac-Tyr-Val-Ala-Asp-CMK 1MG
SDP
Supplier:  CPC Scientific CASP012B
Encompass
Irreversible inhibitor of caspase-1 (ICE) that has also been shown to be a potent therapeutic agent against in vivo liver apoptosis in mice.ONE-LETTER SEQUENCE: Ac-YVAD-CMKMOLECULAR FORMULA: C24H33N4O8Cl1MOLECULAR WEIGHT:541STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [178603-78-6], RESEARCH AREA: ApoptosisREFERENCES: J. B. Schulz et al., J Neurosci, 16, 4696 (1996); N.P.C. Walker et al., Cell 78, 343 (1994)
Catalog No. 50-210-8943
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.