Learn More
ATCC Adult human; ventricular cardio; myocyte/SV-40; gene (AC16)
Supplier:  ATCC CRL3568
Encompass
AC16 cells were derived from primary adult ventricular tissue and can be induced to differentiate into mature cardiomyocytes. This product is an ATCC manufactured and accessioned progeny of PTA-1500 cited in US Pat. No. 7,223,599.
Catalog No. NC2744431
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.