Learn More
Bachem Acetyl-(Asn30,Tyr32)-Calcitonin (8-32) (salmon I) Trifluoroacetate
SDP
Supplier:  Bachem 4034543.1000
Encompass_Preferred
Acetyl-(Asn30,Tyr32)-Calcitonin (8-32) (salmon I) Trifluoroacetate, Ac-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Asn-Thr-Tyr-NH2, 151804-77-2, C127H205N37O40, Mr 2890.25, 1mg. AC187. AC187 is a potent and selective antagonist of amylin-induced hyperglycaemia and hyperlactaemia. It has been used to demonstrate that amylin receptors mediate the anorectic action of salmon calcitonin.
Catalog No. 50-194-6908