Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACACB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114376
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114376 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114376 Supplier Invitrogen™ Supplier No. PA5114376
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat skeletal muscle tissue, mouse skeletal muscle tissue. IHC: rat testis tissue. Flow: HL-60 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Catalyzes the ATP-dependent carboxylation of acetyl-CoA to malonyl-CoA. Carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase. Involved in inhibition of fatty acid and glucose oxidation and enhancement of fat storage. May play a role in regulation of mitochondrial fatty acid oxidation through malonyl-CoA-dependent inhibition of carnitine palmitoyltransferase 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACACB
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene ACACB
Gene Accession No. E9Q4Z2, O00763
Gene Alias Acacb; Acc2; ACCB; ACC-beta; acetyl-CoA carboxylase 2; acetyl-CoA carboxylase beta; acetyl-Coenzyme A carboxylase 2; acetyl-Coenzyme A carboxylase beta; AI597064; AW743042; Biotin carboxylase; HACC275
Gene Symbols ACACB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of human Acetyl Coenzyme A Carboxylase/ACACB (EENPEVAVDCVIYLSQHISPAERAQVVHLLSTMD).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100705, 116719, 32
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.