Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acacetin 98.0+%, TCI America™
SDP

Catalog No. A3013100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3013100MG 100 mg
A301325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3013100MG Supplier TCI America Supplier No. A3013100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 480-44-4
Molecular Formula C16H12O5
Molecular Weight (g/mol) 284.267
MDL Number MFCD00016936
InChI Key DANYIYRPLHHOCZ-UHFFFAOYSA-N
Synonym acacetin, linarigenin, 5,7-dihydroxy-4'-methoxyflavone, acacetine, 4'-methoxyapigenin, buddleoflavonol, linarisenin, akatsetin, apigenin 4'-methyl ether, 5,7-dihydroxy-2-4-methoxyphenyl-4h-chromen-4-one
PubChem CID 5280442
ChEBI CHEBI:15335
IUPAC Name 5,7-dihydroxy-2-(4-methoxyphenyl)chromen-4-one
SMILES COC1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O

Specifications

Melting Point 269°C
Color White-Yellow
Quantity 100 mg
Formula Weight 284.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Acacetin
RTECSNumber DJ3002000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.