Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACADVL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578710
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578710 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578710 Supplier Invitrogen™ Supplier No. PA578710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell lysates, human SiHa whole cell lysates, human A431 whole cell lysates, human U251 whole cell lysates, rat liver tissue lysates, rat heart tissue lysates, mouse liver tissue lysates, mouse heart tissue lysates. IHC: human liver cancer tissue, human ovarian cancer tissue, rat heart tissue. Flow: U251 cell.

The protein encoded by this gene is targeted to the inner mitochondrial membrane where it catalyzes the first step of the mitochondrial fatty acid beta-oxidation pathway. This acyl-Coenzyme A dehydrogenase is specific to long-chain and very-long-chain fatty acids. A deficiency in this gene product reduces myocardial fatty acid beta-oxidation and is associated with cardiomyopathy. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACADVL
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene ACADVL
Gene Accession No. P45953, P49748, P50544
Gene Alias ACAD6; Acadvl; acyl-CoA dehydrogenase, very long chain; acyl-Coenzyme A dehydrogenase, very long chain; LCACD; MVLCAD; Very long chain Acyl-Coa dehydrogenase; very long-chain specific acyl-CoA dehydrogenase, mitochondrial; Vlcad; VLCAD very-long-chain acyl-CoA dehydrogenase
Gene Symbols ACADVL
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human ACADVL (538-576aa RALEQFATVVEAKLIKHKKGIVNEQFLLQRLADGAIDLY).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11370, 25363, 37
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.