Learn More
TARGETMOL CHEMICALS INC ACALABRUTINIB 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3626
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Acalabrutinib (ACP-196) also known as ACP-196 is an orally available inhibitor of Bruton's tyrosine kinase (BTK) with potential antineoplastic activity. Upon administration ACP-196 inhibits the activity of BTK and prevents the activation of the B-cell antigen receptor (BCR) signaling pathway. This prevents both B-cell activation and BTK-mediated activation of downstream survival pathways. This leads to an inhibition of the growth of malignant B cells that overexpress BTK. BTK a member of the src-related BTK/Tec family of cytoplasmic tyrosine kinases is overexpressed in B-cell malignancies; it plays an important role in B lymphocyte development activation signaling proliferation and survival. purity: 99%
Catalog No. 50-258-0193
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.