Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ACAP4 Recombinant Protein Antigen
SDP

Catalog No. NB803523 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB803523 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB803523 Supplier Novus Biologicals™ Supplier No. NBP248550PEP
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1 M Urea, pH 7.4.
Gene ID (Entrez) 55616
Molecular Weight (g/mol) MolecularWeight-theroretical: 25 kDa
Quantity 0.1 mL
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias ARF6 GTPase-activating protein, arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 3, ArfGAP with SH3 domain, ankyrin repeat and PH domain 3, ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 1, centaurin, beta 6, CENTB6, DDEFL1, development and differentiation enhancing factor-like 1, Development and differentiation-enhancing factor-like 1, FLJ20199, Protein up-regulated in liver cancer 1, UPLC1ACAP4, up-regulated in liver cancer 1 (UPLC1)
Gene Symbol ASAP3
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-48550. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mLFor current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com For further blocking peptide related information and a protocol, click here .
Research Category Lipid and Metabolism
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein ACAP4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.