Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACBD6 Monoclonal Antibody (4A9D5E9)
GREENER_CHOICE

Catalog No. PIMA528990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528990 Supplier Invitrogen™ Supplier No. MA528990
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ACBD6.

ACBD6 binds long-chain acyl-coenzyme A molecules with a strong preference for unsaturated C18:1-CoA, lower affinity for unsaturated C20:4-CoA, and very weak affinity for saturated C16:0-CoA. Does not bind fatty acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACBD6
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4A9D5E9
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ACBD6
Gene Accession No. Q9BR61
Gene Alias 0610010G04Rik; 2610100E10Rik; Acbd6; acyl-CoA binding domain containing 6; acyl-CoA-binding domain-containing protein 6; acyl-Coenzyme A binding domain containing 6
Gene Symbols ACBD6
Host Species Mouse
Immunogen Recombinant Human ACBD6 protein (Met1-Ala282).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84320
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.