Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AccelerOme™ TMTpro™ 16plex MS Sample Preparation Kits

Catalog No. PIA50950
Change view
Click to view available options
Quantity:
1 x 16 TMTpro Reactions
2 x 16 TMTpro Reactions
Sufficient For:
10–30 μg samples
30–100 μg samples
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
PIA50950 1 x 16 TMTpro Reactions 30–100 μg samples
PIA50949 2 x 16 TMTpro Reactions 10–30 μg samples
PIA50951 2 x 16 TMTpro Reactions 30–100 μg samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIA50950 Supplier Thermo Scientific™ Supplier No. A50950
Add to Cart
Add to Cart
This item is not returnable. View return policy

TMTpro™ 16plex MS Sample Preparation Kits provide efficient and robust mass spec sample prep with TMTpro peptide labeling for use with the AccelerOme Automated Sample Prep System.

The AccelerOme TMTpro 16plex MS Sample Preparation Kits integrate pre-formulated buffers, MS-grade trypsin/Lys-C enzyme mix, and peptide clean-up cartridges with TMTpro 16plex label reagents for reproducible sample preparation of TMT-labeled peptides. The TMTpro 16plex label reagents have been optimized for use with samples from 10 to 100 μg samples to achieve greater than 95% labeling efficiency.

The Thermo Scientific Accelerome TMTpro 16plex MS Sample Preparation Kits provide a complete solution for efficient and robust proteomics mass spectrometry (MS) sample preparation, including TMT peptide labeling, for use with the AccelerOme Automated Sample Prep System.

TMTpro 16plex MS Sample Preparation Kits features:
Flexible—sample prep and TMT-labeling reagents verified on system using cells, plasma, proteins, and tissue samples for 10 to 100 μg of sample input (kit dependent)
Multiplex—enables concurrent MS analysis of up to 16 samples
Optimized — EasyPep kit chemistry combined with TMTpro 16plex labeling reagents specifically designed for use with the AccelerOme Automated Sample Prep System
Convenient—complete set of ready-to-use proteomic sample prep, clean-up, and TMT-labeling reagents for easy loading onto the instrument
Robust —reagents combined with AccelerOme ecosystem provide reproducible, high-quality TMT-labeled peptides with greater than 95% labeling efficiency

Kit components are single-use and conveniently provided in labeled trays for direct placement on the AccelerOme system. This kit provides a complete reagent solution for automated sample preparation of TMTpro 16plex labeled samples to produce high quality, MS-ready peptides.

The AccelerOme reagent portfolio is an integral part of the AccelerOme Automated system solution to provide robust proteomic mass spec sample preparation. The instrument hardware is built on a proven liquid handling format for reproducible sample processing and optimized on-line μSPE sample clean-up for excellent peptide recovery. TMT label reconstitution and sample labeling and pooling have been incorporated before sample clean-up. Peptide quantitation post-clean-up is achieved through accurate UV spectrophotometric analysis to determine peptide concentrations prior to LC-MS analysis. The complete system (kits + reagents + software) was designed to provide high quality peptides while improving productivity with an easy-to-use automated system. When combined with the industry-leading, high-resolution Orbitrap instruments and software, TMTpro 16plex reagents provide integrated total solutions for quantitative protein expression analysis.

These products are subject to a limited use label license.
 

TRUSTED_SUSTAINABILITY

Specifications

Kit Contents Sample prep module, TMT module
Product Type Reagent Kit
Product Line Thermo Scientific
Sufficient For 30–100 μg samples
Starting Material Protein Samples
Final Product Type TMTpro-labeled peptide sample preparation
Detection Method UV Absorption
Format Kit
Workflow Step Automated mass spec sample prep
Quantity 1 x 16 TMTpro Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.