Learn More
Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers
Description
Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.
Features
- Card access comes with 5 key cards
- Field installed - purchased separately
- Factory installed in the plant with purchase of unit
Specifications
Specifications
| Type | Field-installed |
| For Use With (Equipment) | TSX Universal |
| Includes | Card access w/five key cards |