Learn More
Description
- Round access port with silicone plug
- Choose between 30, 50, or 100 mm plug
- Available on left, right, or top of the chamber
- For use with KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, and KBF PRO 1600
Specifications
Specifications
| For Use With (Equipment) | KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600 |
| Material | Silicone |
| Type | Access Port |
| Ports | 1 on Left Side |
| Port Size | 30 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
