Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Accessories for 1500 Series A2 Class II, Type A2 Biological Safety Cabinet

Catalog No. 13684279
Change view
Click to view available options
Product Type:
Adjustable Stand
Fixed Stand with Rolling Casters
Indented Worksurface Module
Rear Cover Kit
Stainless Steel Padded Armrest
Thimble
Twist-lock Power Cord
Universal Piping
For Use With (Equipment):
3 ft. 1500 A2 Class II BSC
3 ft. 1500 A2 Class II BSC, L, T
3 ft. 1500 A2 Class II BSC, L,T,B
3 ft. 1500 A2 Class II BSC, R, T
3/4/5 ft. 1500 A2 Class II BSC
4 ft. 1500 A2 Class II BSC
4 ft. 1500 A2 Class II BSC, L, T
4 ft. 1500 A2 Class II BSC, L, T, B
4 ft. 1500 A2 Class II BSC, R, T
4 ft. 1500 A2 Class II BSC, R, T, B
5 ft. 1500 A2 Class II BSC
5 ft. 1500 A2 Class II BSC, L, T
5 ft. 1500 A2 Class II BSC, L, T, B
5 ft. 1500 A2 Class II BSC, R, T
5 ft. 1500 A2 Class II BSC, R, T, B
6 ft. 1500 A2 Class II BSC
6 ft. 1500 A2 Class II BSC, L, T
6 ft. 1500 A2 Class II BSC, L, T, B
6 ft. 1500 A2 Class II BSC, R, T
6 ft. 1500 A2 Class II BSC, R, T, B
All heights 1500 A2 Class II BSC
All hts 1500 A2 Class II BSC, L, B
All hts 1500 A2 Class II BSC, R, B
38 product options available for selection
Product selection table with 38 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
13-684-279 Adjustable Stand 4 ft. 1500 A2 Class II BSC
13-684-281 Adjustable Stand 3 ft. 1500 A2 Class II BSC
13-684-282 Adjustable Stand 5 ft. 1500 A2 Class II BSC
13-684-280 Adjustable Stand 6 ft. 1500 A2 Class II BSC
13-684-277 Fixed Stand with Rolling Casters 3 ft. 1500 A2 Class II BSC
13-684-275 Fixed Stand with Rolling Casters 4 ft. 1500 A2 Class II BSC
13-684-278 Fixed Stand with Rolling Casters 5 ft. 1500 A2 Class II BSC
13-684-276 Fixed Stand with Rolling Casters 6 ft. 1500 A2 Class II BSC
13-684-266 Thimble 3/4/5 ft. 1500 A2 Class II BSC
13-684-267 Thimble 6 ft. 1500 A2 Class II BSC
13-684-272 Stainless Steel Padded Armrest All heights 1500 A2 Class II BSC
13-684-268 Rear Cover Kit 3 ft. 1500 A2 Class II BSC
13-684-269 Rear Cover Kit 4 ft. 1500 A2 Class II BSC
13-684-270 Rear Cover Kit 5 ft. 1500 A2 Class II BSC
13-684-271 Rear Cover Kit 6 ft. 1500 A2 Class II BSC
13-684-273 Indented Worksurface Module 3 ft. 1500 A2 Class II BSC
13-684-274 Indented Worksurface Module 4 ft. 1500 A2 Class II BSC
12-090-029 Indented Worksurface Module 5 ft. 1500 A2 Class II BSC
12-090-023 Indented Worksurface Module 6 ft. 1500 A2 Class II BSC
12-090-036 Twist-lock Power Cord All heights 1500 A2 Class II BSC
12-090-030 Universal Piping 3 ft. 1500 A2 Class II BSC
12-090-035 Universal Piping 3 ft. 1500 A2 Class II BSC, R, T
12-090-031 Universal Piping 3 ft. 1500 A2 Class II BSC, L,T,B
12-090-032 Universal Piping 3 ft. 1500 A2 Class II BSC, L, T
12-090-025 Universal Piping 4 ft. 1500 A2 Class II BSC, R, T, B
12-090-027 Universal Piping 4 ft. 1500 A2 Class II BSC, R, T
12-090-021 Universal Piping 4 ft. 1500 A2 Class II BSC, L, T, B
12-090-016 Universal Piping 4 ft. 1500 A2 Class II BSC, L, T
12-090-019 Universal Piping 5 ft. 1500 A2 Class II BSC, R, T, B
12-090-017 Universal Piping 5 ft. 1500 A2 Class II BSC, R, T
12-090-024 Universal Piping 5 ft. 1500 A2 Class II BSC, L, T, B
12-090-018 Universal Piping 5 ft. 1500 A2 Class II BSC, L, T
12-090-026 Universal Piping 6 ft. 1500 A2 Class II BSC, R, T, B
12-090-033 Universal Piping 6 ft. 1500 A2 Class II BSC, R, T
12-090-022 Universal Piping 6 ft. 1500 A2 Class II BSC, L, T, B
12-090-034 Universal Piping 6 ft. 1500 A2 Class II BSC, L, T
12-090-020 Universal Piping All hts 1500 A2 Class II BSC, R, B
12-090-028 Universal Piping All hts 1500 A2 Class II BSC, L, B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 38 options available.
38 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-684-279 Supplier Thermo Scientific™ Supplier No. 1911578
Add to Cart
Add to Cart
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Redefine excellence in your lab with the Thermo Scientific™ 1500 Series Class II, Type A2 Biological Safety Cabinet, offering advanced protection, exceptional user comfort, and convenience.

The Thermo Scientific™ 1500 Series Class II, Type A2 Biological Safety Cabinet combines first-class protection, energy efficiency, and user convenience for today's modern laboratory. Available in nominal widths of 3 ft, 4 ft, 6 ft, and it meets NSF/ANSI Standard 49. Discover how we are delivering enhancements in all 4Cs: Containment, Comfort, Convenience, and Cost Effectiveness.

Containment
  • Dual blower and pressure sensor system enable a higher degree of personal and product protection compared to the traditional, single blower BSC
  • Thermo Scientific™ SmartFlow™ Plus Technology provides autonomous safety features to maintain proper air flow and monitor critical conditions in real time without user interaction
  • Thermo Scientific™ DAVe™ (Digital Airflow Verification) Plus System validates protection by monitoring airflow velocities, detecting, and alerting to any changes
  • H14 HEPA filters remove airborne contaminants with 99.995% efficiency at the most penetrating particle size (MPPS).
Comfort
  • New ergonomically designed window mechanism that offers haptic feedback when moving the window, allowing users to easily locate the optimal window position
  • SmartClean window design allows users to lower the window easily for thorough cleaning of inner surfaces
  • 10° sloped front glass for better comfort and ergonomics
  • Spacious work area with comfortable armrests options
Convenience
  • Newly added, intuitive touch screen provides essential data and status of the cabinet
  • Easily program UV disinfection cycle – customize the date, time, and duration of the UV cycle to fit your schedule
  • Thermo Scientific™ SmartPort™ design provides contained passage of tubing and cables into the work area
Cost Effectiveness
  • Night Stand-by Mode reduced airflow to 30% during low usage periods
  • DC motors offer low energy consumption
Certifications
  • NSF/ANSI 49 Standard, UL, and CE
TRUSTED_SUSTAINABILITY

Specifications

Product Type Adjustable Stand
For Use With (Equipment) 4 ft. 1500 A2 Class II BSC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.