Learn More
Agilent BioTek Accessories for BioStack Microplate Stacker
GSA_VAGREENER_CHOICE
Catalog No. BT1230003 Shop All Agilent Technologies Products
Change view
Click to view available options
Product Type:
10 Plate Stack Set
30 Plate Stack Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
BT1230003 10 Plate Stack Set
BT1230004 30 Plate Stack Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BT1230003 Supplier Agilent BioTek Supplier No. 1230003
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with BioStack4 microplate stacker
  • For use with lidded plates
  • Note: Only for use with Biostack4
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type 10 Plate Stack Set
For Use With (Equipment) Biostack microplate stacker, lidded plates ≤22 mm or unlidded plates ≤17.8 mm