Learn More
Labconco™ Accessories for Labconco™ Fume Hoods, Sash Stop
SDP
Catalog No. 163069 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Basic™ Hood
Catalog No. For Use With (Equipment)
16-306-9 Basic™ Hood
1 options
Catalog No. 16-306-9 Supplier Labconco™ Supplier No. 4965901
Add to Cart
Add to Cart
Sash Stop Kit for use with Labconco Basic 47 and Basic 70 Fume Hoods.
The Sash Stop restricts how far the sash may be opened. This small plastic device may be easily installed on the left side of Labconco Basic 47 and Basic 70 Hoods to restrict sash opening height.
Specifications
For Use With (Equipment) Basic™ Hood
Weight (English) 1 lb.
Weight (Metric) 0.5 Kg
Product Type Sash Stop Kit
WARNING: Cancer - www.P65Warnings.ca.gov