Learn More
Dickson™ Accessories for LCD Display Temperature Data Loggers
Description
Includes
Software: USB downloadable cable; Card reader kit: card reader and removable SD flash card
Requires
Software: Microsoft Windows 98 or newer, 4 MB RAM, 1 free USB port, CD drive
DicksonWare™ Software
- Easy logger setup, data downloads and installation
- For real-time monitoring and data visualization with tables and graphs
- Can reset logger to factory settings
- Transfers data from compatible loggers
- Can be used where data logger cannot be brought back to computer
- For transferring data from card to the DicksonWare software
Specifications
Specifications
| Type | Memory Card |
| Includes | Software, USB downloadable cable, card reader kit, card reader and removable SD flash card |
| For Use With (Application) | Display Temperature and humidity logger, pressure datalogger |