Learn More
MilliporeSigma™ Accessories to Milli-Q™ IQ 7000 Ultrapure Water System
Shop All MilliporeSigma Products
Description
Water purification accessories.
Feed Water Conductivity Cell : Measures the conductivity of water feeding
Foot Pedal: Hands-free dispensing accessory
Tank Level Adapter: For pure feed water stored in lab water storage tank
Water Sensor: Detects water spillage and automatically closes the Milli-Q™ inlet solenoid valve to protect the surroundings from water damage; 120V/60Hz
Order Info
Must be ordered with the Milli-Q™ IQ 7000 Ultrapure Water System. Please contact your Fisher Rep for more details.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Type | Foot Pedal |
| For Use With (Equipment) | Milli-Q IQ 7000 Ultrapure Water System |