Learn More
Eppendorf™ Accessories for New Brunswick S41i Incubator Shaker
Catalog No. 14380017 Shop All Eppendorf North America Products
Change view
Click to view available options
Product Type:
Perforated Shelf Rack
Stacking Kit
Quantity:
1
2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
14-380-017 Stacking Kit 1
14-380-019 Perforated Shelf Rack 1
14-380-018 Perforated Shelf Rack 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-380-017 Supplier Eppendorf™ Supplier No. P06286502
Add to Cart
Add to Cart
Maximize your lab space with the stacking kit for the Eppendorf New Brunswick S41i, allowing you to efficiently stack two units. Enhance your cell culture capabilities with additional shelves designed for static incubation of adherent cells, ensuring optimal growth conditions and streamlined workflow.
Stacking Kit:
  • Allows up to 2 each New Brunswick S41i shakers to be stacked one on top of the other
Additional Perforated Shelves:
  • Can easily be added for static cell culture of adherent cells
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stacking Kit
Description Allows up to 2 each New Brunswick S41i shakers to be stacked one on top of the other
Model New Brunswick S41i Stacking Kit
For Use With (Equipment) New Brunswick S41i
Packaging Quantity 1 Ea.
Quantity 1