Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Argos Technologies™ Accessories and Replacement Parts for E-Vac Aspirating Systems

Argos Technologies Accessories and Replacement Parts for E-Vac Aspirating Systems are for use during solvent waste collection

Supplier:  Cole-Parmer™ 1305040

 View more versions of this product

Catalog No. 03-395-133


Description

Description

Handy replacement and accessory items for aspiration
Specifications

Specifications

Autoclavable
None
15.9 cm
E-Vac Aspirating Systems
Polypropylene
1
Storage Bottle
4 L
Natural
Solvent Waste Collection
4 L Polypropylene Bottle
15.9 cm
Non-sterile
15.9 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.