Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Argos Technologies™ Accessories and Replacement Parts for E-Vac Aspirating Systems

Catalog No. 03395135 Shop All Oakton Products
Change view
Click to view available options
Type:
Barbed Cap
Bottle with Transfer Cap
Cap with Barbed Fittings
Cap with Quick Couplings
Communication Cable
Pipette Adapter
Tubing Set
Vacuum Tubing
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
03-395-135 Cap with Barbed Fittings
03-395-136 Vacuum Tubing
03-395-138 Tubing Set
03-395-134 Cap with Quick Couplings
03-395-139 Communication Cable
03-395-144 Bottle with Transfer Cap
03-395-140 Pipette Adapter
03-395-142 Barbed Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 03-395-135 Supplier Cole-Parmer™ Supplier No. 0439758
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Argos Technologies Accessories and Replacement Parts for E-Vac Aspirating Systems are for use during solvent waste collection

Handy replacement and accessory items for aspiration

Specifications

Autoclavable Autoclavable
Certifications/Compliance None
Color Red
Description Lid for Polypropylene Bottle with barbed tube fittings
Diameter (Metric) Outside 10.2 cm
For Use With (Application) Solvent Waste Collection
For Use With E-Vac Aspirating Systems
Material Polypropylene
Overall Length (Metric) 10.2 cm
Quantity 1
Sterility Non-sterile
Type Cap with Barbed Fittings
Length (Metric) 10.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.