Learn More
Cole-Parmer™ Argos Technologies™ Accessories and Replacement Parts for E-Vac Aspirating Systems
Catalog No. 03395144 Shop All Oakton Products
Change view
Click to view available options
Type:
Barbed Cap
Bottle with Transfer Cap
Cap with Barbed Fittings
Cap with Quick Couplings
Communication Cable
Pipette Adapter
Tubing Set
Vacuum Tubing
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
03-395-144 Bottle with Transfer Cap
03-395-136 Vacuum Tubing
03-395-138 Tubing Set
03-395-134 Cap with Quick Couplings
03-395-135 Cap with Barbed Fittings
03-395-139 Communication Cable
03-395-140 Pipette Adapter
03-395-142 Barbed Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 03-395-144 Supplier Cole-Parmer™ Supplier No. 0439617
Add to Cart
Add to Cart
Argos Technologies Accessories and Replacement Parts for E-Vac Aspirating Systems are for use during solvent waste collection
Handy replacement and accessory items for aspiration
Warranty and Services
None
Specifications
Capacity (Metric) 1 L
Description MiniVac 1L Polystyrene Bottle - Includes Transfer Cap
For Use With (Application) Aspiration
For Use With House vacuum or separate vacuum pump
Material Polystyrene
Model MiniVac
Type Bottle with Transfer Cap