Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers

Catalog No. 502723563
Encompass
Encompass_Preferred
Change view
Click to view available options
Product Type:
Aerosol Cover
Bottle Stand
Bottle Stand, 2 x 2 Configuration
Liquid Filter
Rinse Bottle
Storage Bottle
Storage Bottle Cap
Wash Bottle A
Wash Bottle B
Wash Head (Blank)
For Use With (Equipment):
Wellwash Microplate Strip Washer
Wellwash Versa Microplate Strip Washer
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
50-272-3563 Wash Head (Blank) Wellwash Versa Microplate Strip Washer
50-272-3561 Aerosol Cover Wellwash Microplate Strip Washer
50-272-3562 Wash Bottle A Wellwash Microplate Strip Washer
14430214 Wash Bottle B Wellwash Versa Microplate Strip Washer
14430215 Rinse Bottle Wellwash Versa Microplate Strip Washer
14430218 Aerosol Cover Wellwash Versa Microplate Strip Washer
50-272-3560 Bottle Stand, 2 x 2 Configuration Wellwash Versa Microplate Strip Washer
50-272-3559 Bottle Stand Wellwash Microplate Strip Washer
50-272-3558 Storage Bottle Wellwash Versa Microplate Strip Washer
50-008-33239 Liquid Filter Wellwash Microplate Strip Washer
50-272-3557 Storage Bottle Cap Wellwash Microplate Strip Washer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 50-272-3563 Supplier Thermo Scientific™ Supplier No. N10804
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enhance your Wellwash and Wellwash Versa microplate washers with a range of accessories and replacement parts. Wellwash microplate washers enable reliable ELISA plate washing for 96- or 384-well plates with low residual wash volumes and user-friendly features for optimal convenience and safety.

These accessories and replacement parts are for use with the Wellwash and Wellwash Versa microplate washers. Some are compatible with both models, while others are specifically designed for either the Wellwash or Wellwash Versa microplate washer.

Wellwash microplate strip washers provide reliable and robust plate washing for ELISA applications with low residual wash volumes. Wellwash microplate washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease of use, convenience, and safety.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Wellwash Versa Microplate Strip Washer
Product Type Wash Head (Blank)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.