Learn More
Scott Safety™ Accessory For SEMS II Personnel Accountability System: RFID Programmer
Catalog No. 19823813
Change view
Click to view available options
Product Type:
RFID Card
RFID Card Programmer with Software
Sems II Repeater
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-823-813 Sems II Repeater
19-823-812 RFID Card
19-823-811 RFID Card Programmer with Software
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-823-813 Supplier Scott Safety™ Supplier No. 7012471994
Please contact your local Fisher Scientific sales representative for pricing
For use with the SEMS II system
Scott's SEMS II is a remarkable accountability system that puts the incident commander at every firefighter's side, displaying information vital to personal safety in volatile and dangerous situations, while providing a continuous link of communications to the command station. And with the integrated personnel locator, it provides the peace of mind that a downed or trapped firefighter can be quickly found.
RFID / Personalized Tagging
The system can be personally customized and programmable using RFID identification cards to link a firefighter to a specific Air-Pak™ SCBA. To enter their personal information, each firefighter simply swipes their pre-programmed ID card on the back of the SEMS II console. This information is transmitted to the base command computer while SEMS II is activated. Each card has three programmable fields for firefighter identification that are programmed via an RFID tag writer and proprietary software.
RFID Cards are used to link a firefighter to specific Air-Pak SCBA.
Specifications
Product Type Sems II Repeater