Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCN4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981962
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1962 Supplier Bioss Supplier No. BS12129R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene belongs to the superfamily of acid-sensing ion channels, which are proton-gated, amiloride-sensitive sodium channels. These channels have been implicated in synaptic transmission, pain perception as well as mechanoperception. This gene is predominantly expressed in the pituitary gland, and was considered a candidate for paroxysmal dystonic choreoathetosis (PDC), a movement disorder, however, no correlation was found between mutations in this gene and PDC. Alternative splicing at this locus results in two transcript variants encoding different isoforms.

Specifications

Antigen ACCN4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Asic4
Gene Accession No. Q96FT7
Gene Alias Accn4; acid sensing (proton gated) ion channel family member 4; acid sensing ion channel family member 4; acid sensing ion channel subunit family member 4; acid-sensing (proton-gated) ion channel family member 4; Acid-sensing ion channel 4; Amiloride-sensitive cation channel 4; Amiloride-sensitive cation channel 4, pituitary; amiloride-sensitive cation channel family member 4, pituitary; Asic4; BNAC4; brain sodium channel 4; MGC17248; MGC24860; putative acid-sensing ion channel; Spasic; Spinal chord ASIC
Gene Symbols Asic4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ACCN4/ASIC4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55515
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.