Learn More
KIMBLE™ ACCUFORM™ Round Bottom Disposable Screw Thread Culture Tube
SDP
Catalog No. 01672738 Shop All DWK Life Sciences Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
01-672-738 Autoclavable
1 options
Catalog No. 01-672-738 Supplier KIMBLE™ Supplier No. 7377013100
Add to Cart
Add to Cart
Round bottom disposable screw thread culture tube made of borosilicate glass 5.1.
  • Manufactured from borosilicate glass 5.1
  • 73760 series and 90 series items that end in 'F' have flat bottoms which contribute to stability
  • 73770 series items are produced with a round bottom
  • Modular trays are shrink-wrapped in plastic film
  • Supplied without caps
  • No marking spot
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 8 mL
Length (Metric) 100 mm
Material Borosilicate Glass (5.1)
Shape Cylindrical
Sterility Non-sterile
Autoclavable Autoclavable
Graduated No
Product Type Culture Tube