Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Blank Tags

Catalog No. 17998154 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Black, Orange
Black, Red, White
Blue, White
Message:
DANGER
Notice
WARNING
Material:
Pulp-free Cardstock
RP Plastic
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Message Material
17-998-154 Black, Orange WARNING Pulp-free Cardstock
17-998-152 Black, Red, White DANGER Pulp-free Cardstock
17-998-153 Black, Red, White DANGER RP Plastic
17-998-156 Blue, White Notice Pulp-free Cardstock
17-998-155 Black, Orange WARNING RP Plastic
17-998-157 Blue, White Notice RP Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-998-154 Supplier Accuform Signs Supplier No. MWGT205CTP
Add to Cart
Add to Cart

Use Blank Tags to clearly identify potential hazards in and around your workplace.

Specifications

Color Black, Orange
Diameter (English) Grommet 0.37 in.
Material Pulp-free Cardstock
Message WARNING
For Use With (Application) For indoor and outdoor conditions
Length (Metric) 14.6 cm
Length (English) 5.75 in.
Waterproof Yes
Width (English) 3.25 in.
Width (Metric) 8.25 cm
Dimensions (L x W) 5.75 x 3.25 in.
Diameter (Metric) Hole 0.93 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.