Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Blank Tyvek Tags

Catalog No. 19805256 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-805-256 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-256 Supplier Accuform Signs Supplier No. TBB918P

Write your own convenient message or notice on blank Tyvek™ Tags

Specifications

Color White
Material Pressure-bonded Fiber
Includes Hole with 12 in. galvanized steel wire
Diameter (English) Hole 0.18 in.
Diameter (Metric) Hole 0.45 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.