Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Chemical and Hazardous Safety Labels

Catalog No. 17998262 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
White
Yellow
Displayed Text:
CAUTION Hot
DANGER Flammable
DANGER Hot
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Displayed Text
17-998-262 White DANGER Flammable
17-998-264 White DANGER Hot
17-998-127 Yellow CAUTION Hot
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-998-262 Supplier Accuform Signs Supplier No. LCHL285VSP
Add to Cart
Add to Cart

Colorful, compact safety signs

  • Size: 8.9 × 12.7cm (3.5 × 5 in.)
  • Material: Adhesive vinyl
  • Language: English

Specifications

Material Vinyl
Quantity 5/Pk.
Height (English) 3.5 in.
Height (Metric) 8.9 cm
Max. Temperature (English) 176°F
Min. Temperature (English) -20°F
Thickness 4 mil
Width (English) 5 in.
Width (Metric) 12.7 cm
Temperature Range (Metric) -29°C to 80°C
Temperature Range (English) -20°F to 176°F
Displayed Text DANGER Flammable
Product Type Chemical and hazardous safety label
Color White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.