Learn More
Accuform Signs Circuit Breakers
Catalog No. 19804538 Shop All AccuformNMC Products
Change view
Click to view available options
No. of Poles:
Double
Double/Triple
Single
Single, No Hole
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Poles
19-804-538 Double
19-804-537 Single
19-804-539 Single, No Hole
19-804-541 Double/Triple
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-804-538 Supplier Accuform Signs Supplier No. KDD122
Ensure your employees are safeguarded from unexpected energized breakers with Circuit Breaker Lockouts
Specifications
Product Type Circuit Breaker Lockout
For Use With (Equipment) Fits over breaker two toggles up to 0.62 in. W connected by a cross bar
No. of Poles Double