Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Danger: Confined Space

Catalog No. 17998083 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Aluminum
Plastic
Vinyl
Height (Metric):
17.8 cm
25.4 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (Metric)
17-998-083 Vinyl 17.8 cm
17-998-082 Aluminum 25.4 cm
17-998-081 Plastic 25.4 cm
17-998-085 Aluminum 17.8 cm
17-998-084 Plastic 17.8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-998-083 Supplier Accuform Signs Supplier No. MCSP116VS
Add to Cart
Add to Cart

Colorful, compact safety signs

  • Legend: DANGER CONFINED SPACE
  • Available in a variety of materials and sizes
  • Color: White, Black, Red
  • Language: English

Specifications

Material Vinyl
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage
Height (English) 7 in.
Height (Metric) 17.8 cm
Max. Temperature (English) 146°F
Min. Temperature (English) -20°F
Thickness 4 mil
Width (English) 10 in.
Width (Metric) 25.4 cm
Temperature Range (Metric) -29°C to 63°C
Temperature Range (English) -20°F to 146°F
Languages English
Displayed Text DANGER Confined Space
Product Type Safety Sign
Color Red on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.