Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Electrical Safety Labels

Catalog No. 17998126 Shop All AccuformNMC Products
Change view
Click to view available options
Displayed Text:
DANGER 480 Volts
DANGER Electrical Hazard
DANGER High Voltage
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text
17-998-126 DANGER 480 Volts
17-998-263 DANGER High Voltage
17-998-261 DANGER Electrical Hazard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-998-126 Supplier Accuform Signs Supplier No. LELC164VSP
Add to Cart
Add to Cart

Provide information about potential hazards to keep employees safe when using electrical equipment.

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145
Finish Non-glare
Material Vinyl
Quantity 5/Pk.
For Use With (Application) Provide proper protection in certain areas in order to stay safe on the job or in the for short term outdoor and self sticking applications
Height (English) 3.5 in.
Max. Temperature (English) 176°F
Min. Temperature (English) -20°F
Thickness 4 mil
Width (English) 5 in.
Temperature Range (Metric) -28°C to 80°C
Temperature Range (English) -20°F to 176°F
Displayed Text DANGER 480 Volts
Product Type Electrical Hazard Sign
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.