Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Fluorescent Color Tapes

Catalog No. 19169572 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Fluorescent Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-169-572 Fluorescent Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-169-572 Supplier Accuform Signs Supplier No. PTL410FY
Add to Cart
Add to Cart

Mark a wide variety of areas and objects with easily seen fluorescent flagging tape.

Specifications

Product Type Flagging Tape
Adhesive Type Non-adhesive
Packaging Quantity 1 Roll
Length (Metric) 45.7 m
Thickness 4 mil
Material Vinyl
For Use With (Application) Marking hazards, trees, danger areas, survey markers, boundaries
Length (English) 150 ft.
Width (English) 1.18 in.
Color Fluorescent Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.