Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Hazardous Waste Labels
Ensure the contents in containers are clearly identified and proper precautions are addressed with Container Identification Labels
$60.30 - $80.90
Specifications
Adhesive Type | Self-adhesive |
---|---|
Format | Label |
Material | Vinyl |
Product Type | Hazardous Waste Label |
For Use With (Application) | Hazardous waste |
Catalog Number | Mfr. No. | Display Text | Size (English) | Size (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Display Text | Size (English) | Size (Metric) | Price | Quantity & Availability | |||
19-804-826
|
Accuform Signs
MHZW09EVP |
Hazardous Waste Federal/ State Law | 6 x 6 in. | 15.2 x 15.2 cm |
Pack of 25 for $80.90
|
|
|||
19-804-828
|
Accuform Signs
MHZW15EVP |
Hazardous Waste Handle With Care | 4 x 7 in. | 10.1 x 17.7 cm |
Pack of 25 for $60.30
|
![]() |
|||
19-804-827
|
Accuform Signs
MHZW11EVP |
Non Hazardous Waste Optional Info | 6 x 6 in. | 15.2 x 15.2 cm |
Pack of 25 for $78.40
|
|
|||
19-804-980
|
Accuform Signs
MHZW14EVP |
Non-regulated Waste | 6 x 6 in. | 15.2 x 15.2 cm |
Pack of 25 for $78.40
|
|
|||
Specifications
Self-adhesive | |
Vinyl | |
Hazardous waste | |
-20 to 176°F |
Label | |
Hazardous Waste Label | |
25/Pk. | |
-20°C to +80°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title