Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Safety Signs, Bilingual: No Trespassing

Catalog No. 19804998 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Aluminum
Fiberglass
Displayed Text:
DANGER No Trespassing
Notice No Trespassing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Displayed Text
19-804-998 Fiberglass DANGER No Trespassing
19-804-992 Aluminum Notice No Trespassing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-804-998 Supplier Accuform Signs Supplier No. SBMADM076XF

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

to communicate possible hazards in the workplace or jobsite

Specifications

Material Fiberglass
Quantity 1 Ea.
Description Dura Fiberglass
Height (English) 10 in.
Max. Temperature (English) 140°F
Min. Temperature (English) -20°F
Width (English) 14 in.
Temperature Range (Metric) -28°C to 60°C
Temperature Range (English) -20°F to 140°F
Dimensions (L x W) 10 x 14 in.
Displayed Text DANGER No Trespassing
Product Type Safety Sign
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.